Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKCLPENSA PLIEFANVSQ GILLLLMLKQ HLKNLCGFSD SKIQKYSPSE SAKVYDKAIN RKTGVHFHPK QTLDFLRSDM ANSKITEEVK RSIVKQYLDF KLLMEHLDPD EEEEEGEVSA STNARNKAIT SLLGGGSPKN NTAAETEDDE SDGEDRGGGT SGVRRRRSQR ISQRIT (2523-2686 aa encoded by BC033847)
Probably plays a structural role in chromatin. Involved in sister chromatid cohesion, possibly by interacting with the cohesin complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Delangin; Delangin homolog; DKFZp434L1319; FLJ11203; FLJ12597; FLJ13354; FLJ13648; Nipped-B homolog; Nipped-B-like protein; SCC2 homolog; sister chromatid cohesion 2 homolog
Gene Aliases: CDLS; CDLS1; IDN3; IDN3-B; NIPBL; SCC2
UniProt ID: (Human) Q6KC79, (Mouse) Q6KCD5
Entrez Gene ID: (Human) 25836, (Mouse) 71175, (Rat) 294787
Molecular Function: chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support