Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MLVLRSALT RALASRTLAP QMCSSFATGP RQYDGIFYEF RSYYLKPSKM NEFLENFEKN AHLRTAHSEL VGYWSVEFGG RMNTVFHIWK YDNFAHRTEV RKALAKDKEW QEQFLIPNLA LIDKQESEIT YLVPWCKLEK PPKEGVYELA TFQMKPGGPA LWGDAFKRAV HAHVNLGYTK LVGVFHTEYG ALNRVHVLWW NESADSRAAG RHKSHEDPRV VAAVRESVNY LVSQQNMLLI PTSFSPLK
The NIPSNAP proteins comprise a family of evolutionarily well-conserved proteins with strong sequence similarity to the central portion of a protein encoded by C. elegans chromosome III between a 4-nitrophenylphosphatase (NIP) domain and non-neuronal SNAP25-like protein. NIPSNAP2, a novel gene encoding a protein with tyrosine phosphorylation sites and a transmembrane domain, is co-amplified with EGFR in approximately 40% of glioblastomas, the most common and malignant form of central nervous system tumors. While NIPSNAP3B is highly expressed skeletal muscle, NIPSNAP3A mRNA levels are low. NIPSNAP3A protein is associated with plasma membrane and partially localized in rafts. NIPSNAP proteins have been suggested to be important in vesicular transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: nipsnap homolog 3A; NipSnap3A; NipSnap4; Protein NipSnap homolog 3A; Protein NipSnap homolog 4; Target for Salmonella secreted protein C; TassC
Gene Aliases: 1700054F22Rik; AI429120; HSPC299; NIPSNAP3A; NIPSNAP4; TASSC
UniProt ID: (Human) Q9UFN0
Entrez Gene ID: (Human) 25934, (Rat) 685080, (Mouse) 73398
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support