Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Predicted Reactivity: Mouse (98%), Rat (97%), Zebrafish (82%), Xenopus laevis (85%), Pig (97%), Chicken (91%), Rhesus Monkey (100%), Bovine (97%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NKAP is a transcriptional repressor that associates with the NOTCH (see MIM 190198) corepressor complex and is required for T-cell development.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ22626; NF-kappa-B-activating protein; NF-kappaB activating protein
Gene Aliases: NKAP
UniProt ID: (Human) Q8N5F7
Entrez Gene ID: (Human) 79576
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support