Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Natural killer (NK) cells are lymphocytes that can mediate the lysis of certain tumor cells and virus infected cells without being previously activated. They can also regulate specific humoral and cell mediated immunity. NK cells preferentially express several calcium dependent (C type) lectins, which have been implicated in the regulation of NK cell function. The group, designated KLRC (NKG2) are expressed primarily in natural killer (NK) cells and encodes a family of transmembrane proteins characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C type lectin domain. The KLRC (NKG2) gene family is located within the NK complex, a region that contains several C type lectin genes preferentially expressed on NK cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-lectin type II protein; CD159 antigen-like family member A; CD159 antigen-like family member C; CD159a; CD159c; killer cell lectin-like receptor subfamily C, member 1; killer cell lectin-like receptor subfamily C, member 2; killer cell lectin-like receptor subfamily C, member 3; killer cell lectin-like receptor subfamily C, member 4; natual killer cell group 2-F; natural killer cell lectin; natural killer group protein 2; NK cell receptor A; NK cell receptor C; NK cell receptor E; NK cell receptor F; NKG2-1/B activating NK receptor; NKG2-A/B type II integral membrane protein; NKG2-A/B-activating NK receptor; NKG2-A/NKG2-B type II integral membrane protein; NKG2-C type II integral membrane protein; NKG2-C-activating NK receptor; NKG2-E type II integral membrane protein; NKG2-E-activating NK receptor; NKG2-F type II integral membrane protein; NKG2-F-activating NK receptor
Gene Aliases: CD159A; CD159c; KLRC1; KLRC2; KLRC3; KLRC4; NKG2; NKG2-C; NKG2-E; NKG2-F; NKG2A; NKG2C; NKG2E; NKG2F
UniProt ID: (Human) P26715, (Human) P26717, (Human) Q07444, (Human) O43908
Entrez Gene ID: (Human) 3821, (Human) 3822, (Human) 3823, (Human) 8302
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support