Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: NANDAIGILN NSASFNKMSI EYKYEMMPNR TWRCRVFLQD HCLAEGYGTK KTSKHAAADE ALKILQKTQP TYPSVKSSQC HTGSSPRGSG KKKDIKDLVV YENSSNPVCT LNDTAQFNRM TVEYVYERMT GLRWKCKVIL ESEVIAEAVG VKKTVKYEAA GEAVKTLKKT QPTVINNLKK GAVEDVISRN EIQGRSAEEA YKQQIKEDNI GNQLLRKMGW TGGGLGKSGE GIREPISVKE QHKREGLGLD VERVNKIAKR DIEQIIRNYA RSESHTDLTF SRELTNDERK QIHQIAQKYG LKSKSHGVGH DRYLVVGRKR RKEDLLDQLK QEGQVGHYEL VMPQAN
This gene encodes a transcription factor that interacts with specific negative regulatory elements (NREs) to mediate transcriptional repression of certain NK-kappa-B-responsive genes. The protein localizes predominantly to the nucleolus with a small fraction found in the nucleoplasm and cytoplasm.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ITBA4 protein; NF-kappa-B-repressing factor; NF-kappa-B-repressing factor-like; NFkB-repressing factor; Protein ITBA4; Transcription factor NRF
Gene Aliases: 9430034D17Rik; AW124770; C80287; ITBA4; NKRF; NRF
UniProt ID: (Human) O15226, (Mouse) Q8BY02
Entrez Gene ID: (Rat) 100910054, (Human) 55922, (Mouse) 77286
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support