Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MFPSPALTP TPFSVKDILN LEQQQRSLAA AGELSARLEA TLAPSSCMLA AFKPEAYAGP EAAAPGLPEL RAELGRAPSP AKCASAFPAA PAFYPRAYSD PDPAKDPRAE KKELCALQKA VELEKTEADN AERPRARRRR KPRVLFSQAQ VYELERRFKQ QRYLSAPERD QLASVLKLTS TQVKIWFQNR RYKCKRQRQD QTLELVGLPP PPPPPARRIA VPVLVRDGKP CLGDSAPYAP AYGVGLNPYG YNAYPAYPGY GGAACSPGYS CTAAYPAGPS PAQPATAAAN NNFVNFGVGD LNAVQSPGIP QSNSGVSTLH GIRAW (1-324 aa encoded by BC025711)
This gene encodes a homeobox-containing transcription factor. This transcription factor functions in heart formation and development. Mutations in this gene cause atrial septal defect with atrioventricular conduction defect, and also tetralogy of Fallot, which are both heart malformation diseases. Mutations in this gene can also cause congenital hypothyroidism non-goitrous type 5, a non-autoimmune condition. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cardiac-specific homeobox; cardiac-specific homeobox 1; FLJ52202; FLJ97166; FLJ97195; FLJ97197; FLJ99536; Homeobox protein CSX; Homeobox protein NK-2 homolog E; homeobox protein NKX 2-5; Homeobox protein Nkx-2.5; NK2 transcription factor related locus 5; NK2 transcription factor related, locus 5; NKX 2-5; Nkx-25; tinman paralog
Gene Aliases: CHNG5; CSX; CSX1; HLHS2; NKX2-5; NKX2.5; NKX2E; NKX4-1; VSD3
UniProt ID: (Human) P52952
Entrez Gene ID: (Human) 1482
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support