Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This product is a low-endotoxin, preservative-free formulation. Endotoxin level is <0.01EU/ug.
For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
NKp46 (CD335, NCR1, ) is a cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis. NKp46 consists of two Ig-like domains assembled to leukocyte immunoglobulin-like (LIR) and killer inhibitory receptors (KIR).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-Mar; CD335; hNKp46; Ly-94; Natural cytotoxicity triggering receptor 1; Natural killer cell p46-related protein; natural killer cell receptor; NK-p46
Gene Aliases: NCR1; NKP46
UniProt ID: (Bovine) Q863H2
Entrez Gene ID: (Bovine) 369024
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support