Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence:
MDQPEAPCS STGPRLAVAR ELLLAALEEL SQEQLKRFRH KLRDVGPDGR SIPWGRLERA DAVDLAEQLA QFYGPEPALE VARKTLKRAD ARDVAAQLQE RRLQRLGLGS GTLLSVSEYK KKYREHVLQL HARVKERNAR SVKITKRFTK LLIAPESAAP EEAMGPAEEP EPGRARRSDT HTFNRLFRRD EEGRRPLTVV L
NALP proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins and are thought to be involved in inflammation and reproduction. NALP6, also known as PYPAF5, is highly expressed in granulocytes and T cells and co-localizes with the PYRIN-CARD protein ASC. Co-expression of NALP6 with ASC results in a synergistic activation of NF-kappa-B and activated caspase-1-dependent cytokine processing, suggesting that NALP6 functions in immune cells to coordinate the transduction of pro-inflammatory signals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Angiotensin II/vasopressin receptor; NACHT, leucine rich repeat and PYD containing 6; NACHT, LRR and PYD domains-containing protein 6; NACHT/LRR/pyrin domain-containing protein 6; NLR family, pyrin domain containing 6; Non-angiotensin-vasopressin receptor; Non-AVR; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 6; PYRIN-containing APAF1-like protein 5; PYRIN-containing APAF1-like protein 5-like
Gene Aliases: AI504961; AVR; CLR11.4; NALP6; NAVR; NAVR/AVR; NLRP6; PAN3; PYPAF5
UniProt ID: (Human) P59044, (Mouse) Q8K0L4
Entrez Gene ID: (Human) 171389, (Mouse) 101613
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support