Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTSPQLEWTL QTLLEQLNED ELKSFKSLLW AFPLEDVLQK TPWSEVEEAD GKKLAEILVN TSSENWIRNA TVNILEEMNL TELCKMAKAE MMEDGQVQEI DNPELGDAEE DSELAKPGEK EGWRNSMEKQ SLVWKNTFWQ GDIDNFHDDV TLRNQRFIPF LNPRTPRKLT P
NOD proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins. NALP7, also known as PYPAF3, a member of the related PYRIN-containing APAF1-like proteins (PYPAFs/NALPs) is thought to play a crucial role in cell proliferation. NALP7 has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. It is expressed in numerous tissues including uterus and ovary, with low levels in heart and brain. NALP7 inhibits caspase-1-dependent interleukin-1-b secretion and is a feedback regulator of interleukin-1-b secretion. Defects in the NALP7 gene are known to cause the formation of a hydatidiform mole (HYDM) and reduce the growth of carcinoma cell lines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NACHT, leucine rich repeat and PYD containing 7; NACHT, LRR and PYD containing protein 7; NACHT, LRR and PYD domains-containing protein 7; NLR family, pyrin domain containing 7; Nucleotide-binding oligomerization domain protein 12; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 7; PYRIN-containing APAF1-like protein 3
Gene Aliases: CLR19.4; HYDM; NALP7; NLRP7; NOD12; PAN7; PYPAF3
UniProt ID: (Human) Q8WX94
Entrez Gene ID: (Human) 199713
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support