Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NMB stimulates smooth muscle contraction in a manner similar to that of bombesin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuromedin-B; Neuromedin-B-32; neuromedin-beta
Gene Aliases: 3110023K12Rik; NMB
UniProt ID: (Human) P08949, (Mouse) Q9CR53
Entrez Gene ID: (Human) 4828, (Mouse) 68039
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support