Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: KAVIQVRQK TLHKKTFYYL EQLILKYGMH QNTLRIKEIH DGLDFYYSSK QHAQKMVEFL QCTVPCRYKA SQRLISQDIH SNTYNYKSTF SVEIVPICKD NVVCLSPKLA QSLGNMNQIC VCIRVTSAIH LIDPNTLQVA DIDGSTFWSH PFNSLCHPKQ LEEFIVMECS IVQDIKRAAG AGMISKKHTL GEVWVQKTSE MNTDKQYFCR THLGHLLNPG DLVLGFDLAN CNLNDEHVNK MNSDRVPDVV LIKKSYDRTK RQRRRNWKLK ELARERENMD TDDERQYQDF LEDLEEDEAI RKNVNIYRDS AIPVESDTDD EGAPRISLAE MLEDLHISQD ATGEEGASML T (154-503 aa encoded by BC013317)
NMD3 is a cytoplasmic factor required for a late cytoplasmic assembly step of the 60S subunit but is not a ribosomal protein. A mutation in NMD3 was found to be lethal in the absence of XRN1, which encodes the major cytoplasmic exoribonuclease responsible for mRNA turnover. The NMD3 protein sequence does not contain readily recognizable motifs of known function. However, NMD3 orthologues display an amino-terminal domain containing four repeats of Cx2C, reminiscent of zinc-binding proteins, implicated in nucleic acid binding or protein oligomerization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal export protein NMD3; hNMD3; NMD3 homolog
Gene Aliases: C87860; CGI-07; NMD3; RGD1308468
UniProt ID: (Human) Q96D46, (Mouse) Q99L48
Entrez Gene ID: (Human) 51068, (Mouse) 97112, (Rat) 310512
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support