Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Alomone Labs, LTD
FIGURE: 1 / 2
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
NMDA receptor subtype of glutamate-gated ion channels with reduced single-channel conductance, low calcium permeability and low voltage-dependent sensitivity to magnesium. Mediated by glycine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GluN3B; glutamate [NMDA] receptor subunit 3B; glutamate receptor; Glutamate receptor ionotropic, NMDA 3B; glutamate receptor, ionotropic, NMDA3B; N-methyl-D-aspartate receptor subtype 3B; N-methyl-D-aspartate receptor subunit NR3B; NMDA receptor 4; NMDAR3B; Nr4
Gene Aliases: GluN3B; Grin3b; NMDAR3B; NR3B
UniProt ID: (Mouse) Q91ZU9, (Rat) Q8VHN2
Entrez Gene ID: (Mouse) 170483, (Rat) 170796
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support