Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
NMYC interactor (NMI) encodes a protein that interacts with NMYC and CMYC (two members of the oncogene Myc family), and other transcription factors containing a Zip, HLH, or HLH-Zip motif. The NMI protein also interacts with all STATs except STAT2 and augments STAT-mediated transcription in response to cytokines IL2 and IFN-gamma. The NMI mRNA has low expression levels in all human fetal and adult tissues tested except brain and has high expression in cancer cell line-myeloid leukemias.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N-myc (and STAT) interactor; N-myc and STAT interactor; N-myc interactor; N-myc-interactor; Nmi
Gene Aliases: NMI
UniProt ID: (Human) Q13287
Entrez Gene ID: (Human) 9111
Molecular Function:
transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support