Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Additional Information: This antibody is expected to recognize both reported isoforms (NP_835471.1; NP_001186976.1).
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Catalyzes the formation of NAD(+) from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form; nicotinic acid mononucleotide (NaMN) as substrate with the same efficiency. Can use triazofurin monophosphate (TrMP) as substrate. Can also use GTP and ITP as nucleotide donors. Also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD(+). For the pyrophosphorolytic activity, can use NAD (+), NADH, NAAD, nicotinic acid adenine dinucleotide phosphate (NHD), nicotinamide guanine dinucleotide (NGD) as substrates. Fails to cleave phosphorylated dinucleotides NADP(+), NADPH and NAADP(+). Protects against axonal degeneration following injury.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: naMN adenylyltransferase 1; NaMN adenylyltransferase 3; nicotinamide mononucleotide adenylyltransferase 3; Nicotinamide-nucleotide adenylyltransferase 3; Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 3; nicotinate-nucleotide adenylyltransferase 1; Nicotinate-nucleotide adenylyltransferase 3; NMN adenylyltransferase 3; NMN/NaMN adenylyltransferase 3
Gene Aliases: 4933408N02Rik; Nmnat3; PNAT3
UniProt ID: (Mouse) Q99JR6
Entrez Gene ID: (Mouse) 74080
Molecular Function:
nucleotidyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support