Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: LRLLCPQAF STTVWQFLAV LQEQFGSMAG SNVYLTPPNS QGFAPHYDDI EAFVLQLEGR KLWRVYRPRA PTEELALTSS PNFSQDDLGE PVLQTVLEPG DLLYFPRGFI HQAECQDGVH SLHLTLSTYQ RNTWGDFLEA ILPLAVQAAM EENVEFRRGL PRDFMDYMGA QHSDSKDPRR TAFMEKVRVL VARLGHFAPV DAVADQRAKD FIHDSLPPVL TDRERALSVY GLPIRWEAGE PVNVGAQLTT ETEVHMLQDG IARLVGEGGH LFLYYTVENS RVYHLEEPKC LEIYPQQADA MELLLGSYPE FVRVGDLPCD SVEDQLSLAT TLYDKGLLLT KMPLAL (296-640 aa encoded by BC011350)
Oxygenase that can act as both a histone lysine demethylase and a ribosomal histidine hydroxylase. Specifically demethylates 'Lys-4' (H3K4me) and 'Lys-36' (H3K36me) of histone H3, thereby playing a central role in histone code. Preferentially demethylates trimethylated H3 'Lys-4' (H3K4me3) and monomethylated H3 'Lys-4' (H3K4me1) residues, while it has weaker activity for dimethylated H3 'Lys-36' (H3K36me2). Also catalyzes the hydroxylation of 60S ribosomal protein L8 on 'His-216'. Acts as a regulator of osteoblast differentiation via its interaction with SP7/OSX by demethylating H3K4me and H3K36me, thereby inhibiting SP7/OSX-mediated promoter activation. May also play a role in ribosome biogenesis and in the replication or remodeling of certain heterochromatic region. Participates in MYC-induced transcriptional activation. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L8 histidine hydroxylase; Bifunctional lysine-specific demethylase and histidyl-hydroxylase NO66; FLJ21802; histone lysine demethylase NO66; hsNO66; lysine-specific demethylase NO66; MYC-associated protein with JmjC domain; Nucleolar protein 66; Ribosomal oxygenase 1; Ribosomal oxygenase NO66; ROX; up-regulated in lung cancer 2
Gene Aliases: C14orf169; hsNO66; MAPJD; NO66; RIOX1; ROX; URLC2
UniProt ID: (Human) Q9H6W3
Entrez Gene ID: (Human) 79697
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support