Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MEEQGHSEME IIPSESHPHI QLLKSNRELL VTHIRNTQCL VDNLLKNDYF SAEDAEIVCA CPTQPDKVRK ILDLVQSKGE EVSEFFLYLL QQLADAYVDL RPWLLEIGFS PSLLTQSKVV VNTDPVSRYT QQLRHHLGRD SKFVLCYAQK EELLLEEIYM DTIMELVGFS NESLGSLNSL ACLLDHTTGI LNEQGETIFI LGDAGVGKSM LLQRLQSLWA TGRLDAGVKF FFHFRCRMFS CFKESDRLCL QDLLFKHYCY PERDPEEVFA FLLRFPHVAL FTFDGLDELH SDLDLSRVPD SSCPWEPAHP LVLLANLLSG KLLKGASKLL TARTGIEVPR QFLRKKVLLR GFSPSHLRAY ARRMFPERAL QDRLLSQLEA NPNLCSLCSV PLFCWIIFRC FQHFRAAFEG SPQLPDCTMT LTDVFLLVTE VHLNRMQPSS LVQRNTHSPV ETLHAGRDTL CSLGQVAHRG MEKSLFVFTQ EEVQASGLQE RDMQLGFLRA LPELGPGGDQ QSYEFFHLTL QAFFTAFFLV LDDRVGTQEL LRFFQEWMPP AGAATTSCYP PFLPFQCLQG SGPAREDLFK NKDHFQFTNL FLCGLLSKAK QKLLRHLVPA AALRRKRKAL WAHLFSSLRG YLKSLPRVQV ESFNQVQAMP TFIWMLRCIY ETQSQKVGQL AARGICANYL KLTYCNACSA DCSALSFVLH HFPKRLALDL DNNNLNDYGV RELQPCFSRL TVLRLSVNQI TDGGVKVLSE ELTKYKIVTY LGLYNNQITD VGARYVTKIL DECKGLTHLK LGKNKITSEG GKYLALAVKN SKSISEVGMW GNQVGDEGAK AFAEALRNHP SLTTLSLASN GISTEGGKSL ARALQQNTSL EILWLTQNEL NDEVAESLAE MLKVNQTLKH LWLIQNQITA KGTAQLADAL QSNTGITEIC LNGNLIKPEE AKVYEDEKRI ICF
NOD1 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD1 contains an N-terminal caspase recruitment domain (CARD), a centrally located nucleotide-binding domain (NBD), and ten tandem leucine-rich repeats (LRRs) in its C-terminus. The CARD is involved in apoptotic signaling, and NOD1 activates caspase-9 and NF-kappa-B. LRRs participate in protein-protein interactions, and mutations in the NBD may affect the process of oligomerization and subsequent function of the LRR domain. This protein is an intracellular pattern-recognition receptor (PRR) that initiates inflammation in response to a subset of bacteria through the detection of bacterial diaminopimelic acid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: caspase recruitment domain family, member 4; Caspase recruitment domain-containing protein 4; hNod1; NLR family, CARD domain containing 1; nucleotide-binding oligomerization domain containing 1; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 1; Nucleotide-binding oligomerization domain-containing protein 1
Gene Aliases: CARD4; CLR7.1; NLRC1; NOD1
UniProt ID: (Human) Q9Y239
Entrez Gene ID: (Human) 10392
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support