Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MEEQGHSEME IIPSESHPHI QLLKSNRELL VTHIRNTQCL VDNLLKNDYF SAEDAEIVCA CPTQPDKVRK ILDLVQSKGE EVSEFFLYLL QQL
NOD1 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD1 contains an N-terminal caspase recruitment domain (CARD), a centrally located nucleotide-binding domain (NBD), and ten tandem leucine-rich repeats (LRRs) in its C-terminus. The CARD is involved in apoptotic signaling, and NOD1 activates caspase-9 and NF-kappa-B. LRRs participate in protein-protein interactions, and mutations in the NBD may affect the process of oligomerization and subsequent function of the LRR domain. This protein is an intracellular pattern-recognition receptor (PRR) that initiates inflammation in response to a subset of bacteria through the detection of bacterial diaminopimelic acid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: caspase recruitment domain family, member 4; Caspase recruitment domain-containing protein 4; hNod1; NLR family, CARD domain containing 1; nucleotide-binding oligomerization domain containing 1; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 1; Nucleotide-binding oligomerization domain-containing protein 1
Gene Aliases: CARD4; CLR7.1; NLRC1; NOD1
UniProt ID: (Human) Q9Y239
Entrez Gene ID: (Human) 10392
Molecular Function:
non-motor microtubule binding protein
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support