Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CWPRPPTPPA TNVLLMLYSN LSQEQRQLGG STLLWEAESS WRAQEGQLSW EWGKRHRRHH LPDRSQLCRK VKFQVDFNLI GWGSWIIYPK QYNAYRCEGE
NODAL is an important TGF-beta family protein in cellular differentiation. While constitutively expressed in many developing tissues, NODAL is problematically associated with the differentiation and development of many cancers. The expression of NODAL is regulated by a Notch signaling pathway, which when disrupted contributes to the aggressive behavior of metastatic cancer cells, particularly in melanoma. The abundance of NODAL expression in melanoma can help determine the stage of cancer and metastatic type/progression when tested in IHC. NODAL interactions with Cripto also predispose toward tumor progression. Overexpression of NODAL is also associated with preeclamptic placenta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC138230; Nodal homolog
Gene Aliases: HTX5; NODAL
UniProt ID: (Human) Q96S42
Entrez Gene ID: (Human) 4838
Molecular Function:
growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support