Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVQLRPRASR APASAEAMVD EGQLASEEEE AEHGLLLGQP SSGAAAEPLE EDEEGDDEFD DEAPEELTFA SAQAEAREEE RRVRETVRRD KTLLKEKRKR REELFIEQKK RKLLPDTILE KLTTASQTNI KKSPGKVKEV NLQKKNEDCE KGNDSKKVKV QKVQSVSQNK SYLAVRLKDQ DLRDSRQQAA QAFIHNSLYG PGTNRTTVNK FLSLANKRLP VKRAAVQFLN NAWGIQKKQN AKRFKRRWMV RKMKTKK
NOL7 is encoded by this gene localizes to the nucleolus, where it maintains nucleolar structure and cell growth rates. The encoded protein also functions as a tumor suppressor and regulator of angiogenesis. The RB tumor suppressor gene recruits transcription factors to this gene and positively regulates its expression. Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nucleolar protein 7; nucleolar protein 7, 27kDa; Nucleolar protein of 27 kDa; polyglutamine binding protein 3; retinoic acid repressible protein; U3 small nucleolar RNA-associated protein NOL7; U3 snoRNA-associated protein NOL7
Gene Aliases: C6orf90; dJ223E5.2; NOL7; NOP27; PQBP3; RARG-1
UniProt ID: (Human) Q9UMY1
Entrez Gene ID: (Human) 51406
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support