Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GRKLDPTKE KRGPGRKARK QKGAETELVR FLPAVSDENS KRLSSRARKR AAKRRLGSVE APKTNKSPEA KPLPGKLPKG ISAGAVQTAG KKGPQSLFNA PRGKKRPAPG SDEEEEEEDS EEDGMVNHGD LWGSEDDADT VDDYGADSNS EDEEEGEALL PIERAARKQK (2-170 aa encoded by BC000656)
NOP2 is a member of the unc-84 homolog family and encodes a nuclear nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC117384; MGC149287; MGC149288; NOL1/NOP2/Sun domain family, member 1; NOP2 nucleolar protein homolog; nop2p; Nucleolar protein 1; nucleolar protein 1, 120 kDa; nucleolar protein 1, 120kDa; Nucleolar protein 2 homolog; Probable 28S rRNA (cytosine(4447)-C(5))-methyltransferase; probable 28S rRNA (cytosine-C(5))-methyltransferase; Proliferating-cell nucleolar antigen p120; Proliferation-associated nucleolar protein p120; putative ribosomal RNA methyltransferase NOP2
Gene Aliases: 120kDa; A530002O17; NOL1; NOP120; NOP2; NSUN1; p120
UniProt ID: (Human) B3KPD6
Entrez Gene ID: (Human) 4839, (Mouse) 110109, (Rat) 314969
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support