Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MMAIAPNVTV MVGELVGARL IAHAGSLLNL AKHAASTVQI LGAEKALFRA LKSRRDTPKY GLIYHASLVG QTSPKHKGKI SRMLAAKTVL AIRYDAFGED SSSAMGVENR AKLEARLRTL EDRGIRKISG TGKALAKTEK YEHKSEVKTY DPSGDSTLPT CSKKRKIEQV DKEDEITEKK AKKAKIKVKV EEEEEEKVAE EEETSVKKKK KRGKKKHIKE EPLSEEEPCT STAIASPEKK KKKKKKRENE D
Nop5p, or Nop58p, is localized in the nucleolus and required for pre-18S rRNA processing in Saccharomyces cerevisiae.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NOP58 ribonucleoprotein homolog; Nucleolar protein 5; Nucleolar protein 58; nucleolar protein NOP5/NOP58
Gene Aliases: HSPC120; NOL5; NOP5; NOP5/NOP58; NOP58
UniProt ID: (Human) Q9Y2X3
Entrez Gene ID: (Human) 51602
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support