Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPSKTKYNLV DDGHDLRIPL HNEDAFQHGI CFEAKYVGSL DVPRPNSRVE IVAAMRRIRY EFKAKNIKKK KVSIMVSVDG VKVILKKKKK LLLLQKKEWT WDESKMLVMQ DPIYRIFYVS HDSQDLKIFS YIARDGASNI FRCNVFKSKK KSQAMRIVRT VGQAFEVCHK LSLQHTQQNA DGQEDGESER NSNSSGDPGR QLTGAERAST ATAEETDIDA VEVPLPGNDV LEFSRGVTDL DAVGKEGGSH TGSKVSHPQE PMLTASPRML LPSSSSKPPG LGTETPLSTH HQMQLLQQLL QQQQQQTQVA VAQVHLLKDQ LAAEAAARLE AQARVHQLLL QNKDMLQHIS LLVKQVQELE LKLSGQNAMG SQDSLLEITF RSGALPVLCD PTTPKPEDLH SPPLGAGLAD FAHPAGSPLG RRDCLVKLEC FRFLPPEDTP PPAQGEALLG GLELIKFRES GIASEYESNT DESEERDSWS QEELPRLLNV LQRQELGDGL DDEIAV
This gene encodes a cytosolic protein that binds to the signaling molecule, neuronal nitric oxide synthase (nNOS). This protein has a C-terminal PDZ-binding domain that mediates interactions with nNOS and an N-terminal phosphotyrosine binding (PTB) domain that binds to the small monomeric G protein, Dexras1. Studies of the related mouse and rat proteins have shown that this protein functions as an adapter protein linking nNOS to specific targets, such as Dexras1 and the synapsins. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-terminal PDZ domain ligand of neuronal nitric oxide synthase (CAPON); C-terminal PDZ ligand of neuronal nitric oxide synthase protein; Carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase protein; ligand of neuronal nitric oxide synthase with carboxyl-terminal PDZ domain; MGC138500; nitric oxide synthase 1 (neuronal) adaptor protein; Nitric oxide synthase 1 adaptor protein
Gene Aliases: 6330408P19Rik; CAPON; KIAA0464; NOS1AP
UniProt ID: (Human) O75052
Entrez Gene ID: (Human) 9722
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support