Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SPCANGGRCT QLPSREAACL CPPGWVGERC QLEDPCHSGP CAGRGVCQSS VVAGTARFSC RCPRGFRGPD CSLPDPCLSS PCAHGARCSV GPDGRFLCSC PPGYQGRSCR
NOTCH3 is the third discovered human homologue of the Drosophilia melanogaster type I membrane protein notch. In Drosophilia, notch interaction (with its cell-bound ligands delta, serrate) establishes an intercellular signalling pathway that plays a key role in neural development. Homologues of the notch-ligands have also been identified in human. NOTCH3 functions as a receptor for membrane-bound ligands Jagged1, Jagged2, and Delta1 to regulate cell-fate determination. Mutations in NOTCH3 have been identified as the underlying cause of cerebral autosomal dominant arteriopathy with subcortical infarcts and leukoencephalopathy (CADASIL).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neurogenic locus notch homolog protein 3; Notch 3; Notch homolog 3
Gene Aliases: CADASIL; CADASIL1; CASIL; IMF2; LMNS; NOTCH3
UniProt ID: (Human) Q9UM47
Entrez Gene ID: (Human) 4854
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support