Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Voltage-gated proton (hydrogen) channels play an important role in cellular defense against acidic stress. NOX1 is a homolog of the catalytic subunit of the superoxide-generating NADPH oxidase of phagocytes, gp91phox. Three splice variants of NOX1 have been identified, NOH-1L, NOH-1S and NOH-1Lv. NOH-1S is a voltage-gated proton channel that participates in the regulation of cellular pH and is blocked by zinc. NOH-1L is a pyridine nucleotide-dependent oxidoreductase that generates superoxide and might conduct H (+) ions as part of its electron transport mechanism, whereas NOH-1S does not contain an electron transport chain. NOX1 have the potential to be effective treatments for a range of ischemic diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GP91 phox homolog; Mitogenic oxidase 1; MOX-1; MOX1; NADH/NADPH mitogenic oxidase subunit P65-MOX; NADPH oxidase 1; NADPH oxidase 1 alpha; NOH-1; NOX-1
Gene Aliases: GP91-2; MOX1; NOH-1; NOH1; Nox-1; Nox1; NOX1a; NOX1alpha
UniProt ID: (Rat) Q9WV87, (Mouse) Q8CIZ9
Entrez Gene ID: (Rat) 114243, (Mouse) 237038
Molecular Function: oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support