Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LHNKFWQENR PDYVNIQLYL SQTDGIQKII GEKYHALNSR LFIGRPRWKL LFDEIAKYNR GKTVGVFCCG PNSLSKTLHK LSNQNNSYGT RFEYNKESFS
Oxygen sensing is essential for homeostasis in all aerobic organisms. A phagocyte-type oxidase, similar to that responsible for the production of large amounts of reactive oxygen species (ROS) in neutrophil granulocytes, with resultant antimicrobial activity, has been postulated to function in the kidney as an oxygen sensor that regulates the synthesis of erythropoietin in the renal cortex. NOX4 has a role as a redox messenger in the activation of intracellular signaling pathways leading (or contributing) to mitochondriogenesis, cell survival, and differentiation in hematopoietic stem cells. Data suggests that NOX4 provides a novel link between the insulin receptor and the generation of cellular reactive oxygen species that enhances insulin signal transduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX-1; KOX1; NADPH oxidase 4; NOX 4; NOX-4; Renal NAD(P)H-oxidase; Superoxide-generating NADPH oxidase 4
Gene Aliases: AI648021; KOX; KOX-1; NOX4; RENOX
UniProt ID: (Human) Q9NPH5, (Rat) Q924V1, (Mouse) Q9JHI8
Entrez Gene ID: (Human) 50507, (Rat) 85431, (Mouse) 50490
Molecular Function:
oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support