Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: NLTTPASTSQ DASQCQPSPD FSHDRQLRLL LSQPIQPMMP GSCDARQPSE VSRTGRQVKY AQSQTVFQNP DAHPANSSSA PMPVLLMGQA VLH
Members of the basic helix-loop-helix-PER-ARNT-SIM (bHLH-PAS) family are transcription factors that contain a bHLH DNA binding domain located amino-terminal to a PAS domain. Neuronal PAS domain protein 2 (NPAS2, also designated PAS 4/MOP4) is a member of the bHLH-PAS family and the PAS superfamily. NPAS2, which maps to chromosome 2p11.2-2q13, is expressed primarily in the neurons during the first week of postnatal development. The pattern of NPAS2 expression temporally matches the development of learning and memory, and spatially matches the frontal association/limbic forebrain pathway. NPAS2 may serve a regulatory role in the development and maintenance of long-term memory, and may be required for the processing of complex sensory information. NPAS2 and MOP3 form a transcriptionally active heterodimer which binds to a CACGTGA-containing DNA element and drives transcription from a linked luciferase reporter gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basic-helix-loop-helix-PAS protein MOP4; bHLHe9; Class E basic helix-loop-helix protein 9; Member of PAS protein 4; member of PAS superfamily 4; Neuronal PAS domain-containing protein 2; Neuronal PAS2; PAS domain-containing protein 4
Gene Aliases: BHLHE9; MOP4; NPAS2; PASD4
UniProt ID: (Human) Q99743
Entrez Gene ID: (Human) 4862
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support