Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
FIGURE: 1 / 2
A 1:100 dilution of MA5-45750 was sufficient for detection of NPAS4 in 20 µg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. |Detects approximately 90kDa.approximately
This antibody was formerly sold as clone S408-79.
NXF is a member of the basic helix-loop-helix-PER (MIM 602260)-ARNT (MIM 126110)-SIM (MIM 600892) (bHLH-PAS) class of transcriptional regulators, which are involved in a wide range of physiologic and developmental events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLH PAS type transcription factor NXF; bHLH-PAS type transcription factor NXF; bHLHe79; Class E basic helix-loop-helix protein 79; HLH-PAS transcription factor NXF; LE-PAS; Limbic-enhanced PAS protein; Neuronal PAS domain-containing protein 4; Neuronal PAS4; PAS domain-containing protein 10; PASD10 and LE-PAS
Gene Aliases: BHLHE79; LE-PAS; NPAS4; NXF; PASD10
UniProt ID: (Human) Q8IUM7, (Rat) Q8CJH6, (Mouse) Q8BGD7
Entrez Gene ID: (Human) 266743, (Rat) 266734, (Mouse) 225872
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support