Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Percent identity with other species by BLAST analysis: Human, Gibbon, Marmoset (100%); Gorilla, Monkey (95%); Bovine, Elephant, Panda, Rabbit, Opossum (85%); Mouse, Rat, Horse (80%).
For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Interacts specifically with a number of opioid ligands. Receptor for neuropeptides B and W, which may be involved in neuroendocrine system regulation, food intake and the organization of other signals. Has a higher affinity for neuropeptide B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 7; G-protein coupled receptor 7; MGC129755; neuropeptide B/W receptor 1; Neuropeptides B/W receptor type 1; opioid-somatostatin-like receptor 7
Gene Aliases: GPR7; NPBWR1
UniProt ID: (Human) P48145
Entrez Gene ID: (Human) 2831
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support