Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
By Western blot, PA1-16817 detects heterogeneously glycosylated NPC1 protein with prominent bands at ~170 and ~220 kDa.
Suggested positive control: human fibroblast cell lysate.
Mutations in the Niemann-Pick disease type C1 (NPC1) gene result in a fatal progressive neurodegenerative disorder characterized by an abnormal sequestration of lipids including cholesterol and glycosphingolipids. The NPC1 protein is a large protein that resides in the limiting membrane of endosomes and lysosomes and mediates intracellular cholesterol trafficking via binding of cholesterol to its N-terminal domain. NPC1 transports low-density lipoproteins to late endosomal/lysosomal compartments where they are hydrolyzed and released as free cholesterol. NPC1, in addition to FTO, MC4R, and PTER has recently been shown to be a new risk loci for early-onset and morbid adult obesity in European populations.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ98532; Niemann Pick type C1; Niemann-Pick C disease protein; Niemann-Pick C1 protein; Niemann-Pick disease, type C1; Niemann-Pick type C1; Niemann-Pick type C1 protein; NPC intracellular cholesterol transporter 1; sphingomyelinosis; truncated Niemann-Pick C1
Gene Aliases: A430089E03Rik; C85354; Cdig2; D18Ertd139e; D18Ertd723e; I79_014078; lcsd; nmf164; NPC; NPC1; spm
UniProt ID: (Pig) P56941, (Human) O15118, (Mouse) O35604
Entrez Gene ID: (Pig) 397591, (Human) 4864, (Mouse) 18145, (Chinese hamster) 100689424, (Rat) 266732
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support