Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Immunogen sequence: MGTASSLVSP AGGEVIEDTY GAGGGEACEI PVEVKPKARL LRNSFRRGAG AAAGAGPGSL PRGVGAGGLL GASFKSTGSS VPELEYAAAE YERLRKEYEI FRVSKNQELL SMGRREAKLD TENKRLRAEL; Positive Samples: 293T, Mouse liver; Cellular Location: Cell projection, cilium
Required for normal ciliary development and function. Inhibits disheveled-1-induced canonical Wnt-signaling activity and may also play a role in the control of non-canonical Wnt signaling which regulates planar cell polarity. Probably acts as a molecular switch between different Wnt signaling pathways. Required for proper convergent extension cell movements.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cilia and flagella associated protein 31; Meckel syndrome, type 7; Nephrocystin-3
Gene Aliases: 3632410F03Rik; AI550417; C230078J01; CFAP31; D330020E01Rik; KIAA2000; MKS7; NPH3; NPHP3; pcy; RHPD; RHPD1; SLSN3
UniProt ID: (Human) Q7Z494, (Mouse) Q7TNH6
Entrez Gene ID: (Human) 27031, (Mouse) 74025
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support