Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Control: K562, U87-MG, SK-N-SH
Predicted Reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Bovine (100%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The function of the encoded protein is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: -14 gene protein; Aag; Alpha-globin regulatory element-containing gene protein; alphaRE-14; conserved gene telomeric to alpha globin cluster; GATOR1 complex protein NPRL3; HS-26; HS40; Nitrogen permease regulator 3-like protein; NPR3-like; NPR3-like, GATOR1 complex subunit; Phg; Protein CGTHBA; Prox1; RP23-177M10.6
Gene Aliases: C16orf35; CGTHBA; HS-40; MARE; NPR3; NPRL3; RMD11
UniProt ID: (Human) Q12980
Entrez Gene ID: (Human) 8131
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support