Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Recommended positive controls: A431, HeLa, HepG2, mouse heart, rat heart.
Predicted reactivity: Rhesus Monkey (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NQO1 is a member of the NAD(P)H dehydrogenase (quinone) family and a cytoplasmic 2-electron reductase. This FAD-binding protein forms homodimers and reduces quinones to hydroquinones. NQO1's enzymatic activity prevents the one electron reduction of quinones that results in the production of radical species. Altered expression of the protein has been seen in many tumors and is also associated with Alzheimer's disease (AD).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Azoreductase; Diaphorase (NADH/NADPH); diaphorase (NADH/NADPH) (cytochrome b-5 reductase); Diaphorase 4; diaphorase 4 (NADH/NADPH); diaphorase-4; dioxin-inducible 1; DT-diaphorase; DTD; Menadione reductase; NAD(P)H dehydrogenase (quinone); NAD(P)H dehydrogenase [quinone] 1; NAD(P)H dehydrogenase, quinone 1; NAD(P)H menadione oxidoreductase 1, dioxin inducible; NAD(P)H:menadione oxidoreductase; NAD(P)H:menadione oxidoreductase 1; NAD(P)H:Quinone acceptor oxidoreductase type 1; NAD(P)H:quinone oxidoreductase 1; NAD(P)H:quinone oxireductase; NQ01; Phylloquinone reductase; QR1; Quinone reductase 1
Gene Aliases: AV001255; DHQU; DIA4; DTD; Nmo-1; Nmo1; NMOR1; NMORI; NQO1; Ox-1; Ox1; QR1
UniProt ID: (Human) Q86UK1, (Mouse) Q64669, (Rat) P05982
Entrez Gene ID: (Human) 1728, (Mouse) 18104, (Rat) 24314
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support