Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAGENHQWQG SILYNMLMSA KQTRAAPEAP ETRLVDQCWG CSCGDEPGVG REGLLGGRNV ALLYRCCFCG KDHPRQGSIL YSMLTSAKQT YAAPKAPEAT LGPCWGCSCG SDPGVGRAGL PGGRPVALLY RCCFCGEDHP RQGSILYSLL TSSKQTHVAP AAPEARPGGA WWDRSYFAQR PGGKEALPGG RATALLYRCC FCGEDHPQQG STLYCVPTST NQAQAAPEER PRAPWWDTSS GALRPVALKS PQVVCEAASA GLLKTLRFVK YLPCFQVLPL DQQLVLVRNC WASLLMLELA QDRLQFETVE VSEPSMLQKI LTTRRRETGG NEPLPVPTLQ HHLAPPAEAR KVPSASQVQA IKCFLSKCWS LNISTKEYAY LKGTVLFNPD VPGLQCVKYI QGLQWGTQQI LSEHTRMTHQ GPHDRFIELN STLFLLRFIN ANVIAELFFR PIIGTVSMDD MMLEMLCTKI
This gene encodes a protein that contains a DNA-binding domain. The encoded protein acts as a dominant-negative regulator of transcription which is mediated by the retinoic acid receptor. This protein also functions as an anti-testis gene by acting antagonistically to Sry. Mutations in this gene result in both X-linked congenital adrenal hypoplasia and hypogonadotropic hypogonadism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAX 1; DSS-AHC critical region on the X chromosome protein 1; nuclear hormone receptor; Nuclear receptor DAX-1; Nuclear receptor subfamily 0 group B member 1; nuclear receptor subfamily 0, group B, member 1; R0B1
Gene Aliases: AHC; AHCH; AHX; DAX-1; DAX1; DSS; GTD; HHG; NR0B1; NROB1; SRXY2
UniProt ID: (Human) P51843, (Mouse) Q61066
Entrez Gene ID: (Human) 190, (Mouse) 11614
Molecular Function:
C4 zinc finger nuclear receptor
DNA-binding transcription factor
zinc finger transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support