Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MSSPTTSSL DTPLPGNGPP QPGAPSSSPT VKEEGPEPWP GGPDPDVPGT DEASSACSTD WVIPDPEEEP ERKRKKGPAP KMLGHELCRV CGDKASGFHY NVLSCEGCKG FFRRSVVRGG ARRYACRGGG TCQMDAFMRR KCQQCRLRKC KEAGMREQCV LSEEQIRKKK IRKQQQQESQ SQSQSPVGPQ GSSSSASGPG ASPGGSEAGS QGSGEGEGVQ LTAAQELMIQ QLVAAQLQCN KRSFSDQPKV TPWPLGADPQ SRDARQQRFA HFTELAIISV QEIVDFAKQV PGFLQLGRED QIALLKASTI EIMLLETARR YNHETECITF LKDFTYSKDD FHRAGLQVEF INPIFEFSRA MRRLGLDDAE YALLIAINIF SADRPNVQEP GRVEALQQPY VEALLSYTRI KRPQDQLRFP RMLMKLVSLR TLSSVHSEQV FALRLQDKKL PPLLSEIWDV HE (1-461 aa encoded by BC047750)
The liver X receptors, LXRA and LXRB, form a subfamily of the nuclear receptor superfamily and are key regulators of macrophage function, controlling transcriptional programs involved in lipid homeostasis and inflammation. The inducible LXRA is highly expressed in liver, adrenal gland, intestine, adipose tissue, macrophages, lung, and kidney, whereas LXRB is ubiquitously expressed. Ligand-activated LXRs form obligate heterodimers with retinoid X receptors and regulate expression of target genes containing LXR response elements.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: liver X nuclear receptor beta; Liver X receptor beta; LX receptor beta; nuclear orphan receptor LXR-beta; Nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1, group H, member 2; Oxysterols receptor LXR-beta; steroid hormone-nuclear receptor NER; Ubiquitously-expressed nuclear receptor
Gene Aliases: LXR-b; LXRB; NER; NER-I; NR1H2; RIP15; UNR
UniProt ID: (Human) P55055
Entrez Gene ID: (Human) 7376, (Pig) 100525290
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support