Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MCPVDKAHR NQCQACRLKK CLQAGMNQDA VQNERQPRST AQVHLDSMES NTESRPESLV APPAPAGRSP RGPTPMSAAR ALGHHFMASL ITAETCAKLE PEDADENIDV TSNDPEFPSS PYSSSSPCGL DSIHETSARL LFMAVKWAKN LPVFSSLPFR DQVILLEEAW SELFLLGAIQ WSLPLDSCPL LAPPEASAAG GAQGRLTLAS METRVLQETI SRFRALAVDP TEFACMKALV LFKPETRGLK DPEHVEALQD QSQVMLSQHS KAHHPSQPVR FGKLLLLLPS LRFITAERIE LLFFRKTIGN TPMEKLLCDM FKN (1-322 aa encoded by BC041421)
NR2E3, a NR2 Hepatocyte NF4-Like Receptor, has been suggested to affect cone cell proliferation in eye development. It has been shown to have a role in determining photoreceptor phenotype during human retinogenesis. In RD7 mice, a deletion in NR2E3 results in hereditary retinal dysplasia and degeneration. A homozygous missense mutation in NR2E3 has been linked to retinitis pigmentosa in Crypto-Jews of Belmonte. Two alternatively spliced isoforms have been documented. NR2E3 is expressed in the retina. ESTs have been isolated from cancerous human lung, colon, and ovary, and from normal human eye.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC138414; MGC138416; Nuclear receptor subfamily 2 group E member 3; Photoreceptor-specific nuclear receptor; Retina-specific nuclear receptor; retinal degeneration 7; RP11-295F4.5; taR-5
Gene Aliases: A930035N01Rik; Nr2e3; PNR; rd7; RNR
UniProt ID: (Mouse) Q9QXZ7
Entrez Gene ID: (Pig) 102159757, (Mouse) 23958
Molecular Function:
C4 zinc finger nuclear receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support