Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PLNGHCYLS GYISLLLRAE PYPTSRYGSQ CMQPNNIMGI ENICELAARL LFSAVEWARN IPFFPDLQIT DQVSLLRLTW SELFVLNAAQ CSMPLHVAPL LAAAGLHASP MSADRVVAFM DHIRIFQEQV EKLKALHVDS AEYSCLKAIV LFTSDACGLS DAAHIESLQE KSQCALEEYV RSQYPNQPSR FGKLLLRLPS LRTVSSSVIE QLFFVRLVGK TPIETLIRDM LLSGSSFNWP YMSIQ (178-421 aa encoded by BC004154)
The protein encoded by this gene is a nuclear hormone receptor and transcriptional regulator. The encoded protein acts as a homodimer and binds to 5'-AGGTCA-3' repeats. Defects in this gene are a cause of Bosch-Boonstra optic atrophy syndrome (BBOAS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: avian erythroblastic leukemia viral (v-erb-a) oncogene homolog-like 3; chicken ovalbumin upstream promoter-transcription factor I; COUP transcription factor 1; COUP transcription factor I; COUP-TF I; COUP-TF1; EAR-3; Nuclear receptor subfamily 2 group F member 1; nuclear receptor subfamily 2, group F, member 1; transcription factor COUP 1 (chicken ovalbumin upstream promoter 1, v-erb-a homolog-like 3); V-erbA-related protein 3
Gene Aliases: BBOAS; BBSOAS; COUP-TF1; COUP-TFI; COUPTFA; EAR-3; EAR3; ERBAL3; NR2F1; NR2F2; SVP44; TCFCOUP1; TFCOUP1
UniProt ID: (Human) P10589, (Mouse) Q60632
Entrez Gene ID: (Human) 7025, (Rat) 81808, (Mouse) 13865
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support