Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: RMPPTQPTHG QFALTNGDPL NCHSYLSGYI SLLLRAEPYP TSRFGSQCMQ PNNIMGIENI CELAARMLFS AVEWARNIPF FPDLQITD
This gene encodes a member of the steroid thyroid hormone superfamily of nuclear receptors. The encoded protein is a ligand inducible transcription factor that is involved in the regulation of many different genes. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor related protein 1; Apolipoprotein A-I regulatory protein 1; apolipoprotein AI regulatory protein 1; ARP-1; chicken ovalbumin; chicken ovalbumin upstream promoter transcription factor 2; chicken ovalbumin upstream promoter-transcription factor I; COUP transcription factor 2; COUP transcription factor II; COUP-TF II; COUP-TF2; MGC117452; Nuclear receptor subfamily 2 group F member 2; nuclear receptor subfamily 2, group F, member 2
Gene Aliases: ARP1; CHTD4; COUPTFB; COUPTFII; NF-E3; NR2F1; NR2F2; SVP40; TFCOUP2
UniProt ID: (Human) P24468
Entrez Gene ID: (Human) 7026
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support