Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSQAELSTCS APQTQRIFQE AVRKGNTQEL QSLLQNMTNC EFNVNSFGPE GQTALHQSVI DGNLELVKLL VKFGADIRLA NRDGWSALHI AAFGGHQDIV LYLITKAKYA ASGR
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Notch-regulated ankyrin repeat-containing protein
Gene Aliases: NRARP
UniProt ID: (Human) Q7Z6K4
Entrez Gene ID: (Human) 441478
Molecular Function: protein-binding activity modulator phosphatase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support