Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: REDYICYARF NHTQTIQQKQ PISVKVISVD ELNDTIAANL SDTEFYGAKS SRERPPTFLT PEGNASNKEE LRGNVLSLEC IAEGLPTPII YWAKEDGMLP KNRTVYKNFE KTLQIIHVSE ADSGNYQC
Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
Neuron-glia-related cell adhesion molecule (NRCAM) is a gene located on chromosome 7q31.1, encoding a protein that belongs to the immunoglobulin superfamily of cell adhesion molecules. NRCAM is expressed in the nervous system and plays a vital role in the development and maintenance of neural connections. This adhesion molecule facilitates neuron-neuron and neuron-glia interactions, impacting processes such as axon guidance, synapse formation, and cell signaling, which are crucial for neural development and plasticity. NRCAM is involved in the modulation of signal transduction pathways that influence neuronal growth and repair, contributing to the stabilization of synaptic structures. Alterations in NRCAM expression or function have been linked to neurodevelopmental disorders, including autism spectrum disorders, and certain psychiatric conditions, reflecting its importance in maintaining neural circuit integrity. Research into NRCAM continues to explore its therapeutic potential in neurobiology and its implications in neurological and psychiatric disorders.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hBravo; MGC138845; MGC138846; Neuronal cell adhesion molecule; Neuronal surface protein Bravo; Ng-CAM-related; NgCAM-related cell adhesion molecule; Nr CAM; Nr-CAM
Gene Aliases: KIAA0343; NRCAM
UniProt ID: (Human) Q92823
Entrez Gene ID: (Human) 4897
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support