Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EPPRILTPAN TLYQVIANRP ALLDCAFFGS PLPTIEWFKG AKGSALHEDI YVLHENGTLE IPVAQKDSTG TYTCVARNKL GMAKNEVHLE IKDATWIVKQ PEYAVVQRGS MVSFECKVKH DHTLSLTVLW LKDNRELPSD ERFTVDKDHL VVADVSDDDS GTYTCVANTT LDSVSASAVL SVV
Cell adhesion, ankyrin-binding protein involved in neuron-neuron adhesion. May play a role in the molecular assembly of the nodes of Ranvier.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankyrin-binding cell adhesion molecule NrCAM; hBravo; mBravo; MGC138845; MGC138846; neuron-glia-CAM-related cell adhesion molecule; Neuronal cell adhesion molecule; Neuronal surface protein Bravo; Ng-CAM-related; NgCAM-related cell adhesion molecule; Nr CAM; Nr-CAM; rBravo
Gene Aliases: Bravo; C030017F07Rik; C130076O07Rik; KIAA0343; mKIAA0343; NRCAM
UniProt ID: (Human) Q92823, (Mouse) Q810U4
Entrez Gene ID: (Pig) 100520970, (Human) 4897, (Mouse) 319504, (Rat) 497815
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support