Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat and Orangutan
Cleaves peptide substrates on the N-terminus of arginine residues in dibasic pairs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N-arginine dibasic convertase; Nardilysin; nardilysin (N-arginine dibasic convertase); Nardilysin convertase; NRD convertase
Gene Aliases: 2600011I06Rik; AI875733; hNRD1; hNRD2; NRD-C; NRD1; NRDC
UniProt ID: (Human) O15242, (Mouse) Q8BHG1
Entrez Gene ID: (Human) 4898, (Mouse) 230598
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support