Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 4
This gene is a member of the neuregulin gene family. This gene family encodes ligands for the transmembrane tyrosine kinase receptors ERBB3 and ERBB4 - members of the epidermal growth factor receptor family. Ligand binding activates intracellular signaling cascades and the induction of cellular responses including proliferation, migration, differentiation, and survival or apoptosis. This gene encodes neuregulin 3. NRG3 has been shown to activate the tyrosine phosphorylation of its cognate receptor, ERBB4, and is thought to influence neuroblast proliferation, migration and differentiation by signalling through ERBB4. NRG3 also promotes mammary differentiation during embryogenesis. Linkage studies have implicated this gene as a susceptibility locus for schizophrenia and schizoaffective disorder. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Additional transcript variants have been described but their biological validity has not been verified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:7999; neuregulin 3; neuregulin 3 variant 1; neuregulin 3 variant 10; neuregulin 3 variant 11; neuregulin 3 variant 12; neuregulin 3 variant 13; neuregulin 3 variant 14; neuregulin 3 variant 3; neuregulin 3 variant 4; neuregulin 3 variant 5; neuregulin 3 variant 6; neuregulin 3 variant 7; neuregulin 3 variant 8; neuregulin 3 variant 9; neuregulin-3-like polypeptide; Pro-neuregulin-3, membrane-bound isoform; Pro-NRG3; scaramanga
Gene Aliases: HRG3; NRG3; pro-NRG3; RGD1559678; ska
UniProt ID: (Human) P56975, (Mouse) O35181
Entrez Gene ID: (Human) 10718, (Rat) 498596, (Mouse) 18183
Molecular Function:
growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support