Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MMPSCNRSCS CSRGPSVEDG KWYGVRSYLH LFYEDCAGTA LSDDPEGPPV LCPRRPWPSL CWKISLSSGT LLLLLGVAAL TTGYAVPPKL EGIGEGEFLV LDQRAADYNQ ALGTCRLAGT ALCVAAGVLL AICLFWAMIG WLSQDTKAEP LDPEADSHVE VFGDEPEQQL SPIFRNASGQ SWFSPPASPF GQSSVQTIQP KRDS
Neurensin-2 (NRSN2) is a 204 amino acid multi-pass membrane protein belonging to the VMP family that may be involved in the transport and maintenance of vesicles. Expressed in brain, Neurensin-2 localizes to cell bodies of hippocampus, diagonal band, amygdaloid nucleus, and habenula nucleus, and is a potential tumor suppressor gene and candidate biomarker for long-term survival in patients with hepatocellular carcinoma (HCC). The gene encoding Neurensin-2 maps to human chromosome 20, which comprises approximately 2% of the human genome, contains nearly 63 million bases and encodes over 600 genes, some of which are associated with Creutzfeldt-Jakob disease, amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome. Additionally, chromosome 20 contains a region with numerous genes which are thought important for seminal production and may be potential targets for male contraception.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ23329; Neurensin-2; Neurensin2
Gene Aliases: C20orf98; dJ1103G7.6; NRSN2
UniProt ID: (Human) Q9GZP1
Entrez Gene ID: (Human) 80023
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support