Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAERQEALR EFVAVTGAEE DRARFFLESA GWDLQIALAS FYEDGGDEDI VTISQATPSS VSRGTAPSDN RVTSFRDLIH DQDEDEEEEE GQRFYAGGSE RSGQQIVGPP RKKSPNELVD DLFKGAKEHG AVAVERVTKS PGETSKPRPF AGGGYRLGAA PEEESAYVAG EKRQHSSQDV HVVLKLWKSG FSLDNGELRS YQDPSNAQFL ESIRRGEVPA ELRRLAHGGQ VNLDMEDHRD EDFVKPKGAF KAFTGEGQKL GSTAPQVLST SSPAQQAENE AKASSSILID ESEPTTNIQI RLADGGRLVQ KFNHSHRISD IRLFIVDARP AMAATSFILM TTFPNKELAD ESQTLKEANL LNAVIVQRLT
p47, also known as NSFL1C, UBX1, UBXD10 or UBXN2C, is a 370 amino acid protein that localizes to both the nucleus and the golgi apparatus (specifically to golgi stacks) and contains one SEP domain and one UBX domain. Functioning as part of a ternary complex with VCP (a protein involved in the heterotypic fusion of transport vesicles with their target membranes) and Syntaxin 5, p47 interacts with and reduces the ATPase activity of VCP and is required for the fragmentation of golgi stacks during mitosis and for subsequent reassembly of golgi stacks after mitosis. p47 is subject to phosphorylation during mitosis, which inhibits p47-golgi interaction and is, therefore, required for proper golgi stack formation and cisternal regrowth. Human p47 shares 89% sequence identity with its mouse counterpart, suggesting a conserved role between species. Multiple isoforms of p47 exist due to alternative splicing events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NSFL1 (p97) cofactor (p47); NSFL1 cofactor p47; p97 cofactor p47; RP4-776F14.2; SHP1 homolog; UBX domain-containing protein 2C
Gene Aliases: dJ776F14.1; NSFL1C; P47; UBX1; UBXD10; UBXN2C
UniProt ID: (Human) Q9UNZ2
Entrez Gene ID: (Human) 55968
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support