Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
May play a role in JNK activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Novel SH2-containing protein 1; SH2 domain-containing 3A; SH2 domain-containing protein 3A
Gene Aliases: NSP1; SH2D3A; UNQ175/PRO201
UniProt ID: (Human) Q9BRG2
Entrez Gene ID: (Human) 10045
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support