Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MARSVRVLV DMDGVLADFE AGLLRGFRRR FPEEPHVPLE QRRGFLAREQ YRALRPDLAD KVASVYEAPG FFLDLEPIPG ALDAVREMND LPDTQVFICT SPLLKYHHCV GEKYRWVEQH LGPQFVERII LTRDKTVVLG DLLIDDKDTV RGQEETPSWE HILFTCCHNR HLVLPPTRRR LLSWSDNWRE ILDSKRGAAQ RE (1-201 aa encoded by BC017454)
Pyrimidine 5-prime nucleotidase and type II (UMPH2).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5' nucleotidase, deoxy (pyrimidine), cytosolic type C; 5'(3')-deoxyribonucleotidase, cytosolic type; 5,3-nucleotidase, cytosolic; Cytosolic 5',3'-pyrimidine nucleotidase; Deoxy-5'-nucleotidase 1; dNT-1; epididymis luminal protein 74; uridine 5'-monophosphate phosphohydrolase 2; uridine 5-monophosphate phosphohydrolase 2; uridine 5-prime monophosphate hydrolase 2; uridine monophosphatase 2
Gene Aliases: cdN; DNT; dNT-1; DNT1; HEL74; NT5C; P5N2; PN-I; PN-II; Umph-2; UMPH2
UniProt ID: (Human) Q8TCD5, (Mouse) Q9JM14
Entrez Gene ID: (Human) 30833, (Mouse) 50773, (Rat) 100360582
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support