Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GFFSHLPSQR PFRTLENDEE QEALPSLDKP GWYSQGNAVH LYELLKKMTG KPEPKVVYFG DSMHSDIFPA RHYSNWETVL ILEELR
Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
While the exact function of the protein encoded by this gene is not known, it belongs to the 5'(3')-deoxyribonucleotidase family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-nucleotidase domain-containing protein 1; 5'-nucleotidase, cytosolic II-like 1 protein
Gene Aliases: C6orf200; LP2642; NT5C2L1; NT5DC1
UniProt ID: (Human) Q5TFE4
Entrez Gene ID: (Human) 221294
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support