Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MTMAAAAVV ARGAGARAAT AAALRGGCGT AARGRPCAGP ARPLCTAPGT APDMKRYLWE RYREAKRSTE ELVPSIMSNL LNPDAIFSNN EMSLSDIEI (1-98 aa encoded by BC052293)
NT5DC3 gene ontology annotations related to this gene include 5'-nucleotidase activity; metal ion binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-nucleotidase domain-containing protein 3; GRP94-neighboring nucleotidase
Gene Aliases: GNN; NT5DC3; TU12B1-TY; TU12B1TY
UniProt ID: (Human) Q86UY8
Entrez Gene ID: (Human) 51559
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support