Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: WGRFSKPPED DDANSYENVL ICKQKTTETG AQQEGIGGLC RGDLSLSLAL KTGPTSGLCP SASPEEDEES EDYQNSASIH QWRESRKVMG QLQREASPGP
NTAL (Non-T cell Activation Linker), also known as LAB (Linker for activation of B cells), is a 30 kDa double-palmitoylated transmembrane adaptor protein expressed by B cells, NK cells, mast cells and macrophages. It is a negative regulator of early stages of BCR-dependent B cell signaling and serves as a negative regulator also in mast cells. However, in mast cells, NTAL also contributes to some activation processes, partially overlapping with LAT function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Linker for activation of B-cells; linker for activation of T cells family, member 2; linker for activation of T cells, transmembrane adaptor 2; Linker for activation of T-cells family member 2; Membrane-associated adapter molecule; Non-T-cell activation linker; Williams-Beuren syndrome chromosomal region 15 protein; Williams-Beuren syndrome chromosomal region 5 protein
Gene Aliases: HSPC046; LAB; LAT2; NTAL; WBS15; WBSCR15; WBSCR5; WSCR5
UniProt ID: (Human) Q9GZY6
Entrez Gene ID: (Human) 7462
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support