Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VCGYLFLPW KCLVVVSLRL LFLVPTGVPV RSGDATFPKA MDNVTVRQGE SATLRCTIDN RVTRVAWLNR STILYAGNDK WCLDPRVVLL SNTQTQYSIE IQNVDVYDEG PYTCSVQTDN HPKTSRVHLI VQVSPKIVEI SSDISINEGN NISLTCIATG RPEPTVTWRH ISPKAVGFVS EDEYLEIQGI TREQSGDYEC SASNDVAAPV VRRVKVTVNY PPYISEAKGT GVPVGQKGTL QCEASAVPSA EFQWYKDDKR LIEGKKGVKV ENRPFLSKLI FFNVSEHDYG NYTCVASNKL GHTNASIMLF GETVL (3-316 aa encoded by BC050716)
This gene encodes a member of the IgLON (LAMP, OBCAM, Ntm) family of immunoglobulin (Ig) domain-containing glycosylphosphatidylinositol (GPI)-anchored cell adhesion molecules. The encoded protein may promote neurite outgrowth and adhesion via a homophilic mechanism. This gene is closely linked to a related family member, opioid binding protein/cell adhesion molecule-like (OPCML), on chromosome 11. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hNT; IgLON family member 2; Neurotrimin
Gene Aliases: 6230410L23Rik; B230210G24Rik; HNT; Igdcc2; IGLON2; NT; NTM; NTRI; R75390; UNQ297/PRO337
UniProt ID: (Human) Q9P121
Entrez Gene ID: (Human) 50863, (Mouse) 235106
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support